8046-0349-120 - FAQs
8046-0349-120 - FAQs
View additional product information for 8046-0349-120 - FAQs (8046-0349-120)
1 product FAQs found
Can the eFluor Organic Dyes be frozen?
As with other organic fluorochromes, we do not recommend the eFluor Organic Dyes be frozen.