Single 1.5 mm Comb for OWL Centipede™ Gel System

Catalog number: 810001

 Related applications: Diagnostic Testing

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

• For use with SSP products or any application requiring agarose gel separation
• Multiple formats available to meet various throughput needs
• Includes replacement parts for each unit

Contents & storage

Store at room temperature.

Documents

Manuals & protocols