CaptureSelect™ AVB Sepharose HP Leakage ELISA
Thermo Scientific™

CaptureSelect™ AVB Sepharose HP Leakage ELISA

Thermo Scientific™ CaptureSelect™ AVB Sepharose HP Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using AVB Sepharose HP affinity resin (Cytiva).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81028081010 Assays
8102808011 Assay
Catalog number 810280810
Price (EUR)
9.320,00
Each
Quantity:
10 Assays
Price (EUR)
9.320,00
Each

CaptureSelect AVB Sepharose HP Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using AVB Sepharose HP affinity resin (Cytiva). Even when covalently attached, small amounts of the AVB Sepharose HP affinity ligand can leach off of the chromatography support and co-elute with the virus particles.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect AVB Sepharose HP Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
For Use With (Equipment)Microplate Reader
FormatKit
Quantity10 Assays
TargetAVB Sepharose HP Affinity Ligand
Product LineCaptureSelect
TypeELISA Kit
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C.