CaptureSelect™ Alpha-1 Antitrypsin Select Leakage ELISA
Thermo Scientific™

CaptureSelect™ Alpha-1 Antitrypsin Select Leakage ELISA

Thermo Scientific™ CaptureSelect™ Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva).
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8102870011 Assay
81028701010 Assays
Catalog number 810287001
Price (BRL)
-
Quantity:
1 Assay

CaptureSelect Alpha-1 AntitrypsinSelect Leakage ELISA is designed to detect possible leached ligand of human Alpha-1 antitrypsin (AAT) purified using AAT Select affinity resin (Cytiva). Even when covalently attached, small amounts of the Alpha-1 Antitrypsin Select affinity ligand can leach off of the chromatography support and co-elute with the purified product.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect Alpha-1 Antitrypsin Select Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
For Use With (Equipment)Microplate Reader
FormatKit
Quantity1 Assay
TargetAlpha-1 AntitrypsinSelect Affinity Ligand
Product LineCaptureSelect
TypeELISA Kit
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C.