CaptureSelect™ IgM Ligand Leakage ELISA Kit

Catalog number: 810289010

 Related applications: Antibody Purification | Contaminant & Impurity QC Testing | Protein Purification & Isolation

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

The CaptureSelect™ IgM Ligand Leakage ELISA detects affinity ligand contamination of IgM purified using CaptureSelect™ IgM Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein. This sensitive ELISA detects affinity ligand contamination of less than 1 ng/mL. The assay is designed to minimize interference and to provide accurate quantitation even in the presence of the protein of interest. The CaptureSelect™ IgM Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Assay Type: ELISA
For Use With (Equipment): Microplate Reader
Product Line: CaptureSelect™
Product Size: 10 assays

Contents & storage

Store in freezer (-5 to -30°C).

Documents

Manuals & protocols

Recommended products