CaptureSelect™ HSA Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ HSA Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ HSA Ligand Leakage ELISA is designed to detect possible leached ligand of human serum albumin (HSA) or HSA fusion products purified using CaptureSelect Human Albumin affinity matrix.
Have Questions?
變更視圖buttonViewtableView
產品號碼Quantity
8102970011 Assay
81029701010 Assays
產品號碼 810297001
價格 (TWD)
-
Quantity:
1 Assay

CaptureSelect HSA Ligand Leakage ELISA is designed to detect possible leached ligand of human serum albumin (HSA) or HSA fusion products purified using the CaptureSelect Human Albumin affinity matrix. The HSA affinity ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The HSA Ligand Leakage ELISA kit can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
規格
Detection Limit1 ng/mL
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Shipping ConditionDry Ice
TargetCaptureSelect HSA Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
內容物與存放
Store at -20°C.