CaptureSelect™ HSA Ligand Leakage ELISA
Thermo Scientific™
CaptureSelect™ HSA Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ HSA Ligand Leakage ELISA is designed to detect possible leached ligand of human serum albumin (HSA) or HSA fusion products purified using CaptureSelect Human Albumin affinity matrix.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
81029701010 ensayos
8102970011 ensayo
Número de catálogo 810297010
Precio (USD)
-
Cantidad:
10 ensayos
CaptureSelect HSA Ligand Leakage ELISA is designed to detect possible leached ligand of human serum albumin (HSA) or HSA fusion products purified using the CaptureSelect Human Albumin affinity matrix. The HSA affinity ligand, a 13 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The HSA Ligand Leakage ELISA kit can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
FormatoKit
Cantidad10 ensayos
Línea de productosCaptureSelect
TipoELISA Kit
Unit SizeEach
Contenido y almacenamiento
Store in freezer (-5°C to -30°C).