Orion™ ROSS™ Glass Bodied Combination pH Electrode
Actual product may vary
Orion™ ROSS™ Glass Bodied Combination pH Electrode
Orion™ ROSS™ Glass Bodied Combination pH Electrode
Orion™ ROSS™ Glass Bodied Combination pH Electrode
Orion™ ROSS™ Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ ROSS™ Glass Bodied Combination pH Electrode

Capture accurate measurements of a variety of sample types, including solvents, with Thermo Scientific™ Orion™ glass body pH electrodes.
Have Questions?
Catalog NumberConnector Types
8102BNBNC
Catalog number 8102BN
Price (USD)
567.90
Online exclusive
631.00
Save 63.10
Each
Connector Types:
BNC
Price (USD)
567.90
Online exclusive
631.00
Save 63.10
Each
Thermo Scientific™ Orion™ ROSS Refillable Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. They are available in chemically resistant glass bodies and best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries.
 

Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode fill solution

  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase
Specifications
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Diameter (Metric) Cap16 mm
Drift<0.002 pH/day
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter, BNC connector
IncludesOne 60mL bottle of Orion ROSS filling solution and spout cap
Internal ReferenceROSS
Length (English)4.72 in.
Length (English) Cable39.37 in.
Length (Metric)120 mm
Length (Metric) Cable1 m
Precision0.01
Storage RequirementsShort-term Storage (up to one week): Soak the electrode in ROSS' pH electrode storage solution, Cat. No. 810001, or standard pH electrode storage solution, Cat. No. 910001. For enhanced electrode performance, use ROSS' pH electrode storage solution. Long-term Storage (more than one week): Fill the reference chamber and securely cover the fill hole. Cover the sensing bulb and reference junction with the protective cap, sleeve or storage bottle containing a few drops of storage solution. Before returning the electrode to use, prepare it as a new electrode.
Temperature (English) Operating32°F to 212°F
Connector TypesBNC
JunctionCeramic
MaterialGlass Body
Temperature (Metric) Operating0°C to 100°C
TypeCombination pH Electrode, BNC connector
pH Range0 to 14
Unit SizeEach