CaptureSelect™ IX Select Leakage ELISA
Thermo Scientific™

CaptureSelect™ IX Select Leakage ELISA

Thermo Scientific™ CaptureSelect™ IXSelect Leakage ELISA is designed to detect possible leached ligand of human Factor IX purified using IXSelect affinity resin (Cytiva).
Catalog NumberQuantity
8103000011 Assay
81030001010 Assays
Catalog number 810300001
Price (JPY)
153,100
Each
Quantity:
1 Assay

CaptureSelect IXSelect Leakage ELISA is designed to detect possible leached ligand of human Factor IX purified using IXSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the IXSelect affinity ligand can leach off of the chromatography support and co-elute with the purified product.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect IXSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
For Use With (Equipment)Microplate Reader
FormatKit
Quantity1 Assay
TargetIXSelect Affinity Ligand
Product LineCaptureSelect
TypeELISA Kit
Unit SizeEach
Contents & Storage
Store at -5°C to -30°C.