CaptureSelect™ Lambda Fab Select Leakage ELISA
Thermo Scientific™

CaptureSelect™ Lambda Fab Select Leakage ELISA

Thermo Scientific™ CaptureSelect™ LambdaFabSelect Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using LambdaFabSelect affinity resin (Cytiva).
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
81030801010 ensayos
8103080011 ensayo
Número de catálogo 810308010
Precio (CLP)
-
Cantidad:
10 ensayos

CaptureSelect LambdaFabSelect Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a lambda light chain that have been purified using LambdaFabSelect affinity resin (Cytiva). Even when covalently attached, small amounts of the LambdaFabSelect affinity ligand can leach off of the chromatography support and co-elute with the antibody.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect LambdaFabSelect Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Especificaciones
ensayoELISA
Límite de detección1 ng/mL
Para utilizar con (equipo)Microplate Reader
FormatoKit
Línea de productosCaptureSelect
Tipo de productoLigand Leakage ELISA
Cantidad10 ensayos
Condiciones de envíoDry Ice
ObjetivoLambdaFabSelect Affinity Ligand
Método de detecciónAbsorbance at 450 nm
Formato del kit de inmunoensayoSandwich ELISA Kit
Tipo de muestraLigand Leachate
Especies dianaNot Applicable
Unit SizeEach
Contenido y almacenamiento
Store at -20°C.