CaptureSelect™ IgG-CH1 Ligand Leakage ELISA
Thermo Scientific™
CaptureSelect™ IgG-CH1 Ligand Leakage ELISA
Thermo Scientific™ CaptureSelect™ IgG-CH1 Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect IgG-CH1 Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
81032001010 Assays
8103200011 Assay
Catalog number 810320010
Price (EUR)
9.310,00
Each
Quantity:
10 Assays
Price (EUR)
9.310,00
Each
CaptureSelect IgG-CH1 Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect IgG-CH1 Affinity Matrix. The ligand, a 14 kD recombinant protein, is coupled by multipoint attachment through stable amide bonds to give high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect IgG-CH1 Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
AssayELISA
For Use With (Equipment)Microplate Reader
FormatKit
Product LineCaptureSelect
Product TypeELISA Kit
Quantity10 Assays
Unit SizeEach
Contents & Storage
Store in freezer (-5°C to -30°C).