CaptureSelect™ KappaXP Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ KappaXP Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ KappaXP Ligand Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a kappa light chain that have been purified using CaptureSelect KappaXP Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8103212011 Assay
81032121010 Assays
Catalog number 810321201
Price (USD)
-
Quantity:
1 Assay

CaptureSelect KappaXP Ligand Leakage ELISA is designed to detect possible leached ligand of human Fab fragments or Ig's with a kappa light chain that have been purified using CaptureSelect KappaXP Affinity Matrix. The ligand, a 13-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect KappaXP Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Limit1 ng/mL
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Shipping ConditionDry Ice
TargetCaptureSelect KappaXP Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.