CaptureSelect™ CH1-XL Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ CH1-XL Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8103462011 Assay
81034621010 Assays
Catalog number 810346201
Price (BRL)
-
Quantity:
1 Assay

CaptureSelect CH1-XL Ligand Leakage ELISA Kit is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect CH1-XL Affinity Matrix. The ligand, a 14-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect CH1-XL Ligand Leakage ELISA Kit can be used as a tool to aid in development of an optimal purification process and in routine quality control of in-process streams and final product.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection Limit1 ng/mL
FormatIndividual components
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Shipping ConditionDry Ice
TargetCaptureSelect CH1-XL Affinity Ligand
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Target SpeciesNot Applicable
Unit SizeEach
Contents & Storage
Store at -20°C.