CaptureSelect™ IgE Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ IgE Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ IgE Ligand Leakage ELISA is designed to detect possible leached ligand of IgE purified using CaptureSelect IgE Affinity Matrix.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
8103542011 Assay
81035421010 Assays
Catalog number 810354201
Price (MXN)
-
Quantity:
1 Assay

CaptureSelect IgE affinity ligand is a 13-kD recombinant protein, expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process, small amounts of affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect IgE Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Specifications
Detection Limit1 ng/mL
FormatIndividual components
Ligand TypeCaptureSelect IgE Affinity Ligand
Product LineCaptureSelect
Product TypeLigand Leakage ELISA
Quantity1 Assay
Sufficient For1 Assay
Detection MethodAbsorbance at 450 nm
Immunoassay Kit FormatSandwich ELISA Kit
Sample TypeLigand Leachate
Unit SizeEach
Contents & Storage
Store at -20°C