CaptureSelect™ FcXP Ligand Leakage ELISA
Thermo Scientific™

CaptureSelect™ FcXP Ligand Leakage ELISA

Thermo Scientific™ CaptureSelect™ FcXP Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect FcXP Affinity Matrix.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoCantidad
81037121010 ensayos x placa de 96 pocillos
8103712011 ensayo x placa de 96 pocillos
Número de catálogo 810371210
Precio (CLP)
-
Cantidad:
10 ensayos x placa de 96 pocillos

CaptureSelect FcXP Ligand Leakage ELISA is designed to detect possible leached ligand of antibodies or antibody fragments purified using CaptureSelect FcXP Affinity Matrix. The FcXP affinity ligand is a 13-kD recombinant protein, expressed in yeast and covalently coupled to a chromatography matrix for high chemical stability and low leakage. During the chromatography process small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the protein.

This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. The CaptureSelect FcXP Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.

Para uso exclusivo en investigación. No apto para uso en procedimientos diagnósticos.
Especificaciones
Límite de detección1 ng/mL
FormatoIndividual components
Tipo de ligandoCaptureSelect FcXP Affinity Ligand
Línea de productosCaptureSelect
Tipo de productoLigand Leakage ELISA
Cantidad10 ensayos x placa de 96 pocillos
Condiciones de envíoDry Ice
Suficiente para10 x 96-well plates
ObjetivoCaptureSelect FcXP Ligand
Método de detecciónAbsorbance at 450 nm
Formato del kit de inmunoensayoSandwich ELISA Kit
Tipo de muestraLigand Leachate
Unit SizeEach
Contenido y almacenamiento
Store at -20°C