Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™
Thermo Scientific™

Orion™ ROSS Ultra™ Epoxy Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesLength (Metric) Cable
8107BNUMDBNC (Waterproof), 8-pin Mini-DIN1 m
8107UWMMDBNC (Waterproof), 8-pin Mini-DIN3 m
8107BNURCABNC (Waterproof), RCA1 m
Catalog number 8107BNUMD
Price (USD)
661.50
Online exclusive
735.00
Save 73.50
Each
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
Length (Metric) Cable:
1 m
Price (USD)
661.50
Online exclusive
735.00
Save 73.50
Each
Thermo Scientific™ Orion™ ROSS Ultra Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra line with a built-in ATC. These products offer a low-maintenance, gel-filled, epoxy body and are best used where minimal maintenance is required. Non-refillable Orion ROSS Ultra Triodes come with an 18 month warranty.
 

Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonomous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mLl each. B and D, 60 mL each

Warranty period

  • 18 months from date of purchase
Specifications
DescriptionOrion ROSS Ultra Low Maintenance pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable
Diameter (Metric)12 mm
Filling Solution TypeNone required
For Use With (Equipment)Orion Star Series Meters
Internal ReferenceROSS
Isopotential Point7
Length (Metric)120 mm
Length (Metric) Cable1 m
Precision0.01
Sample CompatibilityTRIS, sulfides, or proteins
Warranty18 Months
Connector TypesBNC (Waterproof), 8-pin Mini-DIN
JunctionGlass Capillary
MaterialRugged Epoxy Body, Filled with Low Maintenance Gel
Temperature (Metric) Operating0°C to 80°C
TypepH/ATC Combination Electrode
pH Range0 to 14
pH Relative Accuracy0.03 pH for buffers using automatic temperature compensation
Unit SizeEach