Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™
Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™
Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™
Thermo Scientific™

Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesLength (Metric) Cable
8157BNUMDBNC (Waterproof), 8-pin Mini-DIN1 m
8157BNURCABNC (Waterproof), RCA1 m
Catalog number 8157BNUMD
Price (USD)
725.40
Online exclusive
806.00
Save 80.60
Each
Connector Types:
BNC (Waterproof), 8-pin Mini-DIN
Length (Metric) Cable:
1 m
Price (USD)
725.40
Online exclusive
806.00
Save 80.60
Each
Thermo Scientific™ Refillable Orion™ ROSS Ultra Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra™ line with a built-in ATC. Best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Refillable Orion™ ROSS Ultra™ Triodes™ come with a two year warranty.
 

Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode fill solution

  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 ml each. B and D, 60 ml each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 24 months from date of purchase
Specifications
DescriptionOrion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and 8-pin MiniDIN Connector and 1 meter cable
Diameter (Metric)12 mm
Filling Solution TypeROSS electrode 3 M KCl fill solution (Cat. No. 810007)
For Use With (Equipment)Orion Star Series Meters
Internal ReferenceROSS
Isopotential Point7
Length (Metric)120 mm
Length (Metric) Cable1 m
Precision0.01
Sample CompatibilityTRIS, sulfides, or proteins
Warranty2 Years
Connector TypesBNC (Waterproof), 8-pin Mini-DIN
JunctionGlass Fiber
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 100°C
TypepH/ATC Combination Electrode
pH Range0 to 14
pH Relative Accuracy0.03 pH for buffers using automatic temperature compensation
Unit SizeEach