Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™
Actual product may vary
Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™
Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™
Thermo Scientific™

Orion™ ROSS Ultra™ Refillable Epoxy Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnector TypesLength (Metric) Cable
8157BNURCABNC (Waterproof), RCA1 m
8157BNUMDBNC (Waterproof), 8-pin Mini-DIN1 m
Catalog number 8157BNURCA
Price (KRW)
734,000
Each
Connector Types:
BNC (Waterproof), RCA
Length (Metric) Cable:
1 m
Price (KRW)
734,000
Each
Thermo Scientific™ Refillable Orion™ ROSS Ultra Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra™ line with a built-in ATC. Best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Refillable Orion™ ROSS Ultra™ Triodes™ come with a two year warranty.
 
Orion™ ROSS Ultra Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.
 
Solutions required 
Electrode fill solution
810007     ROSS pH electrode fill solution, 5 x 60 mL bottles
 
Electrode storage solution
810001     ROSS pH electrode storage solution, 475 mL bottle
 
pH buffers
910168     pH 1.68 buffer, NIST traceable, 475 mL
910104     pH 4.01 buffer, color coded red, NIST traceable, 475 mL
910105     pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
910686     pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
910107     pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL 
910918     pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
910110     pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
910112     pH 12.46 buffer, NIST traceable, 475 mL
810199     All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH
                 electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode
                 storage bottle
 
pH cleaning solution kit
900020     Cleaning solution A and C, 30 ml each. B and D, 60 ml each
 
Items included with 
810007     ROSS pH electrode fill solution 60 mL 1 bottle
 
Warranty period
24 months from date of purchase
Specifications
DescriptionOrion ROSS Ultra Refillable pH/ATC Triode Combination Electrodes with a BNC and RCA Connector and 1 meter cable
Diameter (Metric)12 mm
Filling Solution TypeROSS electrode 3 M KCl fill solution (Cat. No. 810007)
For Use With (Equipment)Mettler Toledo Five and Seven series pH meters
Internal ReferenceROSS
Isopotential Point7
Length (Metric)120 mm
Length (Metric) Cable1 m
Precision0.01
Warranty2 Years
Connector TypesBNC (Waterproof), RCA
JunctionGlass Fiber
MaterialEpoxy Body
Temperature (Metric) Operating0°C to 100°C
TypepH/ATC Combination Electrode
pH Range0 to 14
pH Relative Accuracy0.03 pH for buffers using automatic temperature compensation
Unit SizeEach