Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode
Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode
Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode
Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode
Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode
Thermo Scientific™

Orion™ ROSS™ Semi-Micro Spear Tip Glass Bodied Combination pH Electrode

Pierce soft and semi-solid samples like cheese, meat, or fruit with a refillable spear tip pH electrode.
Have Questions?
Catalog NumberConnector Types
8163BNWPBNC
Catalog number 8163BNWP
Price (USD)
-
Connector Types:
BNC
Thermo Scientific™ Orion™ ROSS Spear Tip Electrodes provide optimum accuracy and stability while having the additional advantages of a double junction design. Best suited for piercing solid/semi-solid samples and measuring small volume samples.
 

Orion™ ROSS Electrodes provide rapid accuracy and temperature response which goes hand in hand with the excellent stability of this reference system. Compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode fill solution

  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 12 months from date of purchase
Specifications
DescriptionWith BNC waterproof connector
Drift<0.002pH/day
Filling Solution Type3M KCl
For Use With (Equipment)Standard pH Meter
Includes60mL bottle of ROSS electrode fill solution
Internal ReferenceROSS
Length (Metric) Tip20 mm
Precision0.01
Sample Size100 μL
Temperature (English) Operating32°F to 212°F
Tip TypeSpear
Connector TypesBNC
JunctionCeramic
MaterialGlass Body
Temperature (Metric) Operating0°C to 100°C
TypeCombination pH Electrode
pH Range0 to 14
Unit SizeEach