ZyMAX™ Goat Anti-Human IgG (H+L) - AP

Catalog number: 817122

Novex™  Related applications: Secondary Detection

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

Immunogen: Human IgG

Specificity: Human IgG (H+L)

Reactivity: Human IgG (H+L)

Applications: ELISA, Immunohistochemistry (FFPE), Immunofluorescence, Immunoprecipitation, WB

ZyMAX™ antibodies are specifically isolated from antigen-affinity columns using advanced elution protocols, leaving only the highest affinity, antigen-specific antibodies. ZyMAX™ conjugates are prepared with modified cross-linkers to achieve optimal conjugation ratios and stability. Improved purification methods virtually eliminate unconjugated components, giving superior sensitivity and lowest possible levels of background. Enhanced sensitivity, higher titer, lower background -- try them today!
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Reactivity: Human
Target Isotype: IgG
Host Species: Goat
Antibody Fragment: Whole Antibody
Label or Dye: AP (Alkaline Phosphatase)
Product Line: ZyMax™
Product Size: 1 mg
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols