Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.1L) capacity, 230V/50Hz, 12A - FAQs
Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezer, 17 cu. ft. (481.1L) capacity, 230V/50Hz, 12A - FAQs
View additional product information for Forma™ 8600 Series -86°C Ultra-Low Temperature Chest Freezers - FAQs (817CV)