Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Actual product may vary
Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Thermo Scientific™
Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberHoldsQuantity
82006464 (8 x 8 Grid)Each
5958100 (10 x 10 Grid)Each
595949 (7 x 7 Grid)12 Pack
596064 (8 x 8 Grid)12 Pack
621281 (9 x 9 Grid)12 Pack
82001616 (4 x 4 Grid)Each
82002525 (5 x 5 Grid)Each
82004949 (7 x 7 Grid)Each
82008181 (9 x 9 Grid)Each
820100100 (10 x 10 Grid)Each
Catalog number 820064
Price (USD)
5.13
Online exclusive
5.70
Each
Holds:
64 (8 x 8 Grid)
Quantity:
Each
Price (USD)
5.13
Online exclusive
5.70
Each
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
  • Use with fiberboard storage boxes to divide box into grids for vial storage
  • Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Holds64 (8 x 8 Grid)
QuantityEach
Unit SizeEach