AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Optimized for a variety of applications requiring larger per well volume processing.
Have Questions?
Catalog NumberPore SizeVolume (Metric)Material (Membrane)
82470.2 μm2 mLPTFE
Catalog number 8247
Price (MXN)
-
Pore Size:
0.2 μm
Volume (Metric):
2 mL
Material (Membrane):
PTFE
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifications
For Use With (Application)Solvent Filtration
Quantity5/Pk.
Volume (Metric)2 mL
Material (Membrane)PTFE
Pore Size0.2 μm
SterilityNon-sterile
Unit SizeEach