Thermo Scientific™
Citrus Clearing Solvent
 Related applications:
Anatomical Pathology
Replace xylene in both manual and automated system applications with Thermo Scientific™ Citrus Clearing Solvent.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • D-limonene-based solvent has a slight citrus scent.
  • Higher flash point eliminates the need for storage in a flammable safety cabinet.
Documents
Manuals & protocols