Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™
Thermo Scientific™

Orion™ ROSS Ultra™ Glass Bodied pH/ATC Triode™

Accurately measure pH and temperature together in just one electrode, with a built-in temperature sensor.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호커넥터 유형
8302BNUMDBNC 방수, 8핀 MiniDIN, 8-pin Mini-DIN
8302BNURCABNC (Waterproof), RCA
카탈로그 번호 8302BNUMD
제품 가격(KRW)
787,000
온라인 행사
Ends: 31-Dec-2026
925,000
할인액 138,000 (15%)
Each
커넥터 유형:
BNC 방수, 8핀 MiniDIN, 8-pin Mini-DIN
제품 가격(KRW)
787,000
온라인 행사
Ends: 31-Dec-2026
925,000
할인액 138,000 (15%)
Each
Thermo Scientific™ Refillable Orion™ ROSS Ultra™ Triode™ 3-in-1 pH/Automatic Temperature Compensation Electrodes combine the precision and speed of response of the ROSS Ultra™ line with a built-in ATC. Best suited for use in strong acids/bases, pH > 12 or pH < 2, food & cosmetic emulsions, pure water and slurries. Refillable Orion™ ROSS Ultra™ Triodes™ come with a two year warranty.
 

Orion™ ROSS Ultra™ Electrodes offer the best stability and drift free measurements of all our ROSS electrodes while still providing the rapid accuracy and temperature response synonymous with this reference system These are compatible with samples containing organic compounds, Tris buffers, enzymes, proteins, heavy metals, and other compounds.

Solutions required:

Electrode fill solution

  • 810007 ROSS pH electrode fill solution, 5 x 60 mL bottles

Electrode storage solution

  • 810001 ROSS pH electrode storage solution, 475 mL bottle

pH buffers

  • 910168 pH 1.68 buffer, NIST traceable, 475 mL
  • 910104 pH 4.01 buffer, color coded red, NIST traceable, 475 mL
  • 910105 pH 5.00 buffer, color coded orange, NIST traceable, 475 mL
  • 910686 pH 6.86 buffer, DIN standard, NIST traceable, 475 mL
  • 910107 pH 7.00 buffer, color coded yellow, NIST traceable, 475 mL
  • 910918 pH 9.18 buffer, DIN standard, NIST traceable, 475 mL
  • 910110 pH 10.01 buffer, color coded blue, NIST traceable, 475 mL
  • 910112 pH 12.46 buffer, NIST traceable, 475 mL
  • 810199 All-in-One ROSS pH buffer kit, includes pH 4.01, 7.00 and 10.01 buffers, 475ml each ROSS pH electrode storage solution, 475 ml, pH electrode cleaning solution C, 30 ml and 12 mm electrode storage bottle

pH cleaning solution kit

  • 900020 Cleaning solution A and C, 30 mL each. B and D, 60 mL each

Items included with

  • 810007 ROSS pH electrode fill solution 60 mL 1 bottle

Warranty period

  • 24 months from date of purchase
사양
설명BNC 및 8핀 MiniDIN 커넥터와 1미터 케이블을 포함하는 Orion ROSS Ultra 유리 단자 pH/ATC 복합 전극
직경(미터법)12mm
충전 용액 유형ROSS 전극 3 M KCl 충진 용액(Cat. No. 810007)
용도(장비)Thermo Scientific Orion Versa Star, Star A 및 Dual Star 측정기
내부 참조ROSS
등전위 지점7
길이(미터법)120mm
길이(미터법) 케이블1m
정밀도0.01
워런티2년
커넥터 유형BNC 방수, 8핀 MiniDIN, 8-pin Mini-DIN
접합부세라믹
물질화학물질 저항 유리체
작동 온도(미터법) 작동0° ∼ 100°C
유형pH/ATC Combination Electrode
pH 범위0 ∼ 14
pH 상대 정확도자동 온도 보상을 사용하는 완충액 0.03pH
Unit SizeEach