Thermo Scientific™
Richard-Allan Scientific™ Signature Series Flex™ 100, 95, 80
Related applications:
Anatomical Pathology
Achieve brilliant cytoplasmic staining and complete dehydration with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Flex™ 100, 95, 80.
Back to top
  • Patented formulation of methyl and isopropyl alcohol minimizes over-dehydration in smaller biopsies
  • Excellent for lipid extraction allowing for increased paraffin infiltration
  • May be used as a solvent for special stains