Nalgene™ LDPE, Round Carboy with Spigot
Thermo Scientific™

Nalgene™ LDPE, Round Carboy with Spigot

For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Have Questions?
카탈로그 번호 8318-0020
제품 가격(KRW)
-
For Use With: Collect and dispense distilled water, acids and reagents; Closure Material: Polypropylene; Color: White
Features:
  • Molded in graduations of 1 gal. and 5L increments for ease in dispensing
  • Integral shoulder handles that make lifting comfortable for four largest sizes
  • Molded-in spigot boss for strength and leak resistance
  • Leakproof†

Ordering Information:

Supplied with shoulder loop on 1 gal. (3.8L) size, shoulder handles on all other sizes. 1 gal. size is not graduated.

Includes:

Polypropylene spigot and screw closure

Warranty: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com.

사양
용량(영국식 단위)2.6 gal.
마개 재질Polypropylene
마개 크기83B
설명Round Carboys w/Spigot
직경(미터법) 내부 넥64 mm
직경(미터법) 외부250 mm
용도(애플리케이션)Collect and dispense distilled water, acids and reagents
눈금Molded-in graduations in 1 gal. and 5L increments
높이(미터법) 마개 포함389 mm
포함Integral shoulder handles, molded-in spigot boss
케이스당 수6/Cs.
팩당 수1 Ea.
용량(미터법)10 L
색상White
물질LDPE
제품 유형Carboy with Spigot
Unit SizeEA