Thermo Scientific™
Richard-Allan Scientific™ Histoplast Paraffin
Related applications:
Anatomical Pathology
Meet the demanding and versatile needs of the laboratory with Thermo Scientific™ Richard-Allan Scientific™ Histoplast Paraffin.
Back to top
  • Made of paraffin and plastic polymers
  • Delivers excellent processing and yields thin, wrinkle-free sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable 2 lb. bags.