Thermo Scientific™
Richard-Allan Scientific™ Paraffin Type 1, 3, 6, 9
 Related applications:
Anatomical Pathology
Find the perfect option to fit your needs with Thermo Scientific™ Richard-Allan Scientific Paraffin Type 1, 3, 6, 9: Four different blends of highly refined paraffin and polymers, offering a choice of infiltration times and cutting characteristics.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
  • All paraffins melt between 55° and 57°C and are triple filtered to 0.5μm
  • Flake form has a greater surface-to-weight ratio than pellets, resulting in a faster melting time
  • Type 1 contains no polymer and offers rapid infiltration
  • Type 3 contains minimal polymer and offers fast infiltration
  • Type 6 is harder than Type 3 and allows for thinner sections
  • Type 9 has the highest polymer content available and yields extremely thin sections
Ordering Information:
Packaged in convenient, easy-to-open, resealable bags
Documents
Manuals & protocols