Thermo Scientific™
Richard-Allan Scientific™ Signature Series Paraffin
Related applications:
Anatomical Pathology
Minimize compression and obtain rapid infiltration with Thermo Scientific™ Richard-Allan Scientific™ Signature Series Paraffin, a unique blend of polymers and highly refined paraffin with narrow carbon chain distribution.
Back to top
  • Triple-filtered to eliminate impurities
  • Type H allows for ultra-thin sections
  • Type L may reduce infiltration times during processing
  • Both Type L and Type H can be used for infiltration and embedding, and have a melting point of 55° to 57°C
Ordering Information: Packaged in easy-to-open, resealable bags.
test