Raman Organic Chemical Spectral Library
Thermo Scientific™

Raman Organic Chemical Spectral Library

Obtain a large cross-selection of organic chemicals with the Thermo Scientific™ Raman Organic Chemical Spectral Library.
Have Questions?
Número do catálogoType
834-004401Raman Organic Chemical Spectral Library
Número do catálogo 834-004401
Preço (BRL)
-
Type:
Raman Organic Chemical Spectral Library

Obtain a large cross-selection of organic chemicals with the Thermo Scientific™ Raman Organic Chemical Spectral Library, which offers a wide range of functional groups to aid chemical investigations.

Enable confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications with our comprehensive libraries. They feature fully-reviewed, high-quality spectroscopy databases to take the guesswork out of spectral analysis and create greater efficiencies in your lab.

  • All samples were run directly through the unopened brown glass bottles from the Chem Service 0-1000 Mini Stockroom kit using a Thermo Scientific FT-Raman spectrometer.
  • Contains 1,000 spectra collected by using a Thermo Scientific FT-Raman spectrometer
  • Data format: Deresolved
About Spectral Libraries
Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Especificações
For Use With (Application)FT-IR
TypeRaman Organic Chemical Spectral Library
Unit SizeEach