Sigma-Aldrich™ Dyes, Stains and Pigments FT-IR Spectral Library
Thermo Scientific™

Sigma-Aldrich™ Dyes, Stains and Pigments FT-IR Spectral Library

Perform qualitative identification with this material-specific subset of the larger Sigma Biochemical Condensed Phase Library related to Dyes, Stains and Pigments.
Have Questions?
Catalog NumberType
834-006001Dyes, Stains and Pigments FT-IR Spectral Library
Catalog number 834-006001
Price (MXN)
-
Type:
Dyes, Stains and Pigments FT-IR Spectral Library
Perform qualitative identification with this material-specific subset of the larger Sigma Biochemical Condensed Phase Library related to Dyes, Stains and Pigments. The Thermo Scientific™ Sigma Dyes, Stains and Pigments FT-IR Spectral Library is just one of our comprehensive libraries, featuring fully-reviewed, high-quality spectroscopy databases to ensure confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications. Eliminate guesswork in your spectral analysis and create greater efficiencies in your lab.
  • This library is acquired by Sigma-Aldrich Co.™ and examined and processed by Thermo Fisher Scientific. The spectra include compound name, molecular formula, CAS (Chemical Abstract Service) registry number and Sigma catalog number.
  • Contains 628 spectra
  • Data format: Deresolved
About Spectral Libraries

Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification, or at least classification, of the material.
Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeDyes, Stains and Pigments FT-IR Spectral Library
Unit SizeEach