Flavors and Fragrances FT-IR Spectral Library
Thermo Scientific™  Related applications: Lab Automation
Thermo Scientific™ Flavors and Fragrances FT-IR Spectral Library represents a collection of vapor phase chemical compounds of interest to food and cosmetic scientists.
Error loading your content!
  Catalog number Price ({{currency}}) Availability Qty
{{product.sku}}
also known as {{product.formattedSku}} 
{{product.availability.message}} Add To Cart
{{addItemToList.successMessage}} {{addItemToList.listName}}
Back to top
Description
This library contains a wide array of spectra of chemicals which contribute fragrance and flavor to commercial products.
  • Most of the spectra were collected by Aldrich using a GC interface to reduce contamination and decomposition
  • All spectra are on the GRAS (Generally Recognized as Safe) list of US government-approved foods
  • Spectra are complementary to the condensed phase spectra included in the Food Additives Library
  • Contains 667 spectra
  • Data format: High Resolution
Documents
Manuals & protocols