Coatings Technology FT-IR Spectral Library
Actual product may vary
Thermo Scientific™

Coatings Technology FT-IR Spectral Library

Analyze of a wide variety of common coating materials with this valueable spectral library and associated handbook.
Have Questions?
變更視圖buttonViewtableView
產品號碼Type
834-010901MSpectral Library, Member Price
834-010901Spectral Library
產品號碼 834-010901M
價格 (HKD)
-
Type:
Spectral Library, Member Price

Analyze of a wide variety of common coating materials with the Thermo Scientific™ Coatings Technology Library and associated handbook.

Take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Our comprehensive libraries feature fully-reviewed, high-quality spectroscopy databases, enabling confident compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.

The Coatings Technology Library includes spectra of polymers, resins, vehicles, pigments, extenders, acrylic monomers, radiation-curable monomers, fungicides, mildewcides, inhibitors, stabilizers, solvents and other additives. Samples were prepared as capillary films, melts on KBr, cast films on KRS-5 or KBr pellets.
Contains 2,507 spectra of common coating materials
  • Prepared by the Federation of Societies for Coatings Technology
  • Data format: High Resolution (HR)
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification—or at least classification—of the material.
  • Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Note:

Members of the Federation of Societies for Coatings Technology qualify for a 10% discount when purchasing this library. Please be sure to provide your member number when ordering.

Includes:

Revised edition of An Infrared Spectroscopy Atlas for the Coatings Industry (1991). The atlas is the fourth edition of this FSCT publication, dealing with the use of FT-IR in coatings analysis—a 2-volume hardcover book set with valuable reference information on spectroscopy theory, analytical instrumentation, sampling accessories, sample preparation, spectral interpretation and quantitative analysis.
規格
For Use With (Application)FT-IR
TypeSpectral Library, Member Price
Unit SizeEach