Hazardous Chemicals Vapor Phase FTIR Spectral Library
Thermo Scientific™

Hazardous Chemicals Vapor Phase FTIR Spectral Library

Vapor phase spectra of toxic chemicals, pollutants and other contaminants found on the EPA's Toxic Chemical list.
Have Questions?
Catalog NumberType
834-011700Haz Chemicals Vapor Phase FTIR Spectral Library
Catalog number 834-011700
Price (USD)
-
Type:
Haz Chemicals Vapor Phase FTIR Spectral Library

Thermo Scientific™ Hazardous Chemicals Vapor Phase FTIR Spectral Library contains vapor phase spectra of toxic chemicals, pollutants and other contaminants found on the EPA's Toxic Chemical list.

The Hazardous Chemicals Vapor Phase Library contains spectra which are on the US Environmental Protection Agency's Office of Toxic Substances consolidated list of chemicals subject to reporting under Title III of the Superfund Amendments and Reauthorization Act of 1986, a.k.a., the Emergency Planning and Community Right-to-Know Act.
  • Contains 304 spectra
  • Data format: High Resolution
Specifications
For Use With (Application)FT-IR
TypeHaz Chemicals Vapor Phase FTIR Spectral Library
Unit SizeEach