Aldrich™ Aldehydes and Ketones FT-IR Spectral Library
Aldrich™ Aldehydes and Ketones FT-IR Spectral Library
Thermo Scientific™

Aldrich™ Aldehydes and Ketones FT-IR Spectral Library

Confidently identify compounds with the Thermo Scientific™ Aldrich™ Aldehydes and Ketones FT-IR Spectral Library, a subset of Aldrich FT-IR Spectral Library.
Have Questions?
Catalog NumberType
834-019700Aldehydes and Ketones FT-IR Spectral Library
Catalog number 834-019700
Price (EUR)
-
Request A Quote
Type:
Aldehydes and Ketones FT-IR Spectral Library

Confidently identify compounds with the Thermo Scientific™ Aldrich™ Aldehydes and Ketones FT-IR Spectral Library, a subset of Aldrich FT-IR Spectral Library. Our comprehensive libraries take the guesswork out of your spectral analysis and create greater efficiencies in your lab. Fully-reviewed, high-quality spectroscopy databases enable compound identification for a variety of environmental, forensic, pharmaceutical and industrial applications.

The Aldrich Material-Specific FT-IR Library collection represents a wide variety of the most common chemicals in The Aldrich Handbook of Fine Chemicals, divided by similar functional groups. These spectra were assembled from the Aldrich Collections of FT-IR Spectra Editions I or II, and the data has been carefully examined and processed by Thermo Fisher Scientific.
  • Contains 1,311 spectra
  • Data format: High Resolution (HR)
About Spectral Libraries:
  • Spectral library searching has long been an extraordinarily valuable tool in the analytical chemist's toolbox. Our extensive series of spectral libraries includes information specific to application, industry or spectral technique.
  • Spectral library searching based on digital matching of spectral signatures is a very effective way of performing qualitative identification of compounds. Because FT-IR and Raman spectroscopic signatures are based on molecular functional groups, even if the exact compound is not present in the libraries, the list of matching compounds from a spectral library search can often lead to identification—or at least classification—of the material.
  • Thermo Scientific libraries span applications for Hazmat, Forensics, Food, Polymers and Organic Chemistry.
Specifications
For Use With (Application)FT-IR
TypeAldehydes and Ketones FT-IR Spectral Library
Unit SizeEach