Minerals FT-IR Spectral Library
Thermo Scientific™

Minerals FT-IR Spectral Library

A subset of the Inorganics Library that contains minerals from all over the world.
Have Questions?
Catalog NumberType
834-025700Minerals FT-IR Spectral Library
Catalog number 834-025700
Price (MXN)
-
Type:
Minerals FT-IR Spectral Library

Thermo Scientific™ Minerals FT-IR Spectral Library is a subset of the Inorganics Library that contains minerals from all over the world.

  • Contains spectra of 600 minerals Includes information on the mineralogical name, chemical formulas, occurrence and color
  • Data format: High Resolution (HR)
Specifications
For Use With (Application)FT-IR
TypeMinerals FT-IR Spectral Library
Unit SizeEach