Thermo Scientific™
Calibration Validation Carousels
Catalog number: 840-213000
Thermo Scientific™
Calibration Validation Carousels
Catalog number: 840-213000
Thermo Scientific™ Performance Verification Standards, Automated (CVC) will ensure the accuracy and reliability of your data while saving time and money. The CVC provides certified reference materials in a fully automated confirguration for hands-free performance verification of your Thermo Scientific™ spectrophotometer.
Catalog Number
840-213000
Unit Size
Each
Type
Pharmacopia Peformance Validation - EP
Includes
Calibration Validation Carousel with Smart Base
Price (USD)
Full specifications
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypePharmacopia Peformance Validation - EP
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel with Smart Base
Model840-213000
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Routine Performance Validation - NIST Traceable Filters
Includes
Calibration Validation Carousel
Price (USD)
Full specifications
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypeRoutine Performance Validation - NIST Traceable Filters
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel
Model10010701
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Routine Performance Validation - NPL Traceable Filters
Includes
Calibration Validation Carousel with Smart Base
Price (USD)
Full specifications
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypeRoutine Performance Validation - NPL Traceable Filters
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel with Smart Base
Model10010721
Unit SizeEach
Catalog Number
Unit Size
Each
Type
Pharmacopia Peformance Validation - EP
Includes
Calibration Validation Carousel
Price (USD)
Full specifications
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypePharmacopia Peformance Validation - EP
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel
Model840-212800
Unit SizeEach
Showing 4 of 4
Catalog NumberSpecificationsUnit SizeTypeIncludesPrice (USD)
840-213000Full specifications
EachPharmacopia Peformance Validation - EPCalibration Validation Carousel with Smart BaseRequest A Quote
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypePharmacopia Peformance Validation - EP
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel with Smart Base
Model840-213000
Unit SizeEach
10010701Full specifications
EachRoutine Performance Validation - NIST Traceable FiltersCalibration Validation CarouselRequest A Quote
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypeRoutine Performance Validation - NIST Traceable Filters
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel
Model10010701
Unit SizeEach
10010721Full specifications
EachRoutine Performance Validation - NPL Traceable FiltersCalibration Validation Carousel with Smart BaseRequest A Quote
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypeRoutine Performance Validation - NPL Traceable Filters
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel with Smart Base
Model10010721
Unit SizeEach
840-212800Full specifications
EachPharmacopia Peformance Validation - EPCalibration Validation CarouselRequest A Quote
Certifications/ComplianceAll standards contained in the CVC are certified in the Thermo Fisher Scientific ISO/IEC 17025 accredited laboratory traceable to the NIST in the United States or the NPL in the United Kingdom.
DescriptionNIST traceable
TypePharmacopia Peformance Validation - EP
For Use With (Equipment)Evolution 201, 220, 260 Bio and 300
IncludesCalibration Validation Carousel
Model840-212800
Unit SizeEach
Showing 4 of 4
  • Simplify qualification procedures by automating performance verification of UV-Visible spectrophotometers
  • Each CVC has a unique serial number matched automatically to the corresponding data files containing standard values and instrument specifications
  • Generated result reports are suitable for printing or archival purposes
  • Included standards are traceable to either NIST™ primary standards or fundamental physical standards
  • CVCs are available in standard or pharmacopeia compliant configurations
Comprehensive Performance Verification Testing
  • Wavelength Accuracy and Repeatability
  • Photometric Accuracy and Repeatability
  • Stray Light
  • Resolution
  • Photometric Noise
  • Photometric Stability or Drift
  • Baseline Flatness
Save Time, Reduce Errors
  • Eliminates need to copy standard values from certificates
  • Eliminates need to analyze standards individually
  • Eliminates need to match standards to data
  • Eliminates need for manual calculations
  • Eliminates need for manual transcription and reporting of results
Ordering Information:
Unit is shipped in a temperature-protected plastic case for optimal protection during storage. Retain this case and packaging to use when sending the unit for periodic recalibration. Calibration Validation Carousels can be used with Thermo Scientific™ INSIGHT™ and INSIGHT Security, Thermo Scientific™ VISIONpro™ and VISIONsecurity™. Pharmacopeia-compliant CVCs are compatible with the Thermo Scientific™ Evolution 300, 260 Bio, 220 and 201 UV-Visible spectrophotometers.
Figures