NanoDrop™ Lite Printer Accessory
Thermo Scientific™

NanoDrop™ Lite Printer Accessory

Use an optional printer with your NanoDrop™ Lite Spectrophotometer. Print data on freezer-compatible labels.
Have Questions?
Catalog NumberFor Use With (Equipment)
840-218900NanoDrop Lite Spectrophotometer
Catalog number 840-218900
Price (USD)
-
For Use With (Equipment):
NanoDrop Lite Spectrophotometer

Use an optional printer with your NanoDrop™ Lite Spectrophotometer. Print data on freezer-compatible labels.

Print sample data on a cryogenic label

  • Sample number
  • Date
  • Time of Measurement
  • Sample Name
  • Assay
  • Concentration
  • Purity Ratio

For use with the NanoDrop Lite Spectrophotometer

Specifications
DescriptionPrinter Accessory for the NanoDrop Lite
For Use With (Equipment)NanoDrop Lite Spectrophotometer
Unit SizeEach