Specifications
For Use With (Application)Replace your spectrophotometer lid and hinge assembly yourself if it is broken through abuse.
For Use With (Equipment)GENESYS 30 Visible Spectrophotometer
Includes1 lid and hinge assembly
Storage RequirementsAmbient
Warranty2 Years
TypeReplacement Compartment Lid
Unit SizeEach