GENESYS™ 140/150 Vis/UV-Vis Spectrophotometers

Catalog number: 840-300000-1Y

GENESYS™ 140/150 Vis/UV-Vis Spectrophotometers

Catalog number: 840-300000-1Y
Perform quantitative measurements in routine industrial QA/QC or university chemistry labs with the  automated and network-capable Thermo Scientific™ GENESYS™ 150 UV-Visible and GENESYS 140 Vis Spectrophotometers. Optimized for usability and performance, this platform features a high-resolution color touchscreen, optional Wi-Fi networking, and a rugged design built for repetitive, heavy use environments.  Regarded worldwide for reliability, accuracy and reproducibility, GENESYS spectrophotometers meet today’s expectations for advanced technology in a compact, robust package.
 
Catalog Number
840-300000-1Y
Unit Size
Each
Description
GENESYS 150 UV-Vis Spectrophotometer with 1 year Extended Warranty
Optical Design
Dual beam
Spectral Bandwidth
2 nm
Wavelength Range
190 nm - 1100 nm
Price (USD)
6,450.00
Your Price:
Availability
***
Full specifications
Quantity
TypeGENESYS 150 UV-Vis Spectrophotometer with 1 year Extended Warranty
Wavelength Range190 nm - 1100 nm
DescriptionGENESYS 150 UV-Vis Spectrophotometer with 1 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth2 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 150 UV-Visible Spectrophotometer
Optical Design
Dual beam
Spectral Bandwidth
2 nm
Wavelength Range
190 nm – 1100 nm
Price (USD)
6,450.00
Your Price:
Availability
***
Full specifications
Quantity
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range190 nm – 1100 nm
Baseline Flatness±0.002A
Compartments
Accommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 150 UV-Visible Spectrophotometer
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0005A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
Lamp Xenon Flash Lamp (>5 years typical, 3 years guaranteed)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display-3A to +5A
Photometric Range–2A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth2 nm
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedSlow, medium and fast - up to 1600 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 150 UV-Vis Spectrophotometer with EU Power Cable
Optical Design
Dual beam
Spectral Bandwidth
2 nm
Wavelength Range
190 nm – 1100 nm
Price (USD)
Availability
***
Full specifications
Quantity
-
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A±0.004A at 1.0A±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range190 nm – 1100 nm
Baseline Flatness±0.002A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 150 UV-Vis Spectrophotometer with EU Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0005A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–6.0 Hz
KeypadTouchscreen
LampXenon Flash Lamp (>5 years typical, 3 years guaranteed)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display -3A to +5A
Photometric Range–2A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth2 nm
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedSlow, medium and fast - up to 1600 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 150 UV-Vis Spectrophotometer with UK Power Cable
Optical Design
Dual beam
Spectral Bandwidth
2 nm
Wavelength Range
190 nm – 1100 nm
Price (USD)
Availability
***
Full specifications
Quantity
-
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range190 nm – 1100 nm
Baseline Flatness±0.002A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 150 UV-Vis Spectrophotometer with UK Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0005A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampXenon Flash Lamp (>5 years typical, 3 years guaranteed)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display–3A to +5A
Photometric Range–2A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth2 nm
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 140 Visible Spectrophotometer with EU Power Cable
Optical Design
Dual beam
Spectral Bandwidth
5 nm
Wavelength Range
325 nm – 1100 nm
Price (USD)
Availability
***
Full specifications
Quantity
-
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-in. color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range325 nm – 1100 nm
Baseline Flatness±0.005A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 140 Visible Spectrophotometer with EU Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5cm
Drift(At 500 nm after 1 hour warmup) <0.0010A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampTungsten-Halogen (>1000 hours)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display–3A to +5A
Photometric Range–3A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth5 nm
Stray Light<0.05%T at 340 nm and 400 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability<±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 140 Visible Spectrophotometer
Optical Design
Dual beam
Spectral Bandwidth
5 nm
Wavelength Range
325 nm – 1100 nm
Price (USD)
4,580.00
Your Price:
Availability
***
Full specifications
Quantity
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument
±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range325 nm – 1100 nm
Baseline Flatness±0.005A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 140 Visible Spectrophotometer
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0010A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampTungsten-Halogen (>1000 hours)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display-3A to +5A
Photometric Range–3A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth5 nm
Stray Light<0.05%T at 340 nm and 400 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
 
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 140 Visible Spectrophotometer with UK Power Cable
Optical Design
Dual beam
Spectral Bandwidth
5 nm
Wavelength Range
325 nm – 1100 nm
Price (USD)
Availability
***
Full specifications
Quantity
-
Photometric Accuracy Including Standard Tolerance±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-in. color touchscreen, tiltable, high definition, 800 × 1280 pixels
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range325 nm – 1100 nm
Baseline Flatness±0.005A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 140 Visible Spectrophotometer with UK Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5cm
Drift(At 500 nm after 1 hour warmup) <0.0010A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampTungsten-Halogen (>1000 hours)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display–3A to +5A
Photometric Range–3A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth5 nm
Stray Light<0.05%T at 340 nm and 400 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability<±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 140 Vis Spectrophotometer with 1 year Extended Warranty
Optical Design
Dual beam
Spectral Bandwidth
5 nm
Wavelength Range
325 nm – 1100 nm
Price (USD)
4,580.00
Your Price:
Availability
***
Full specifications
Quantity
TypeGENESYS 140 Vis Spectrophotometer with 1 year Extended Warranty
Wavelength Range325 nm – 1100 nm
DescriptionGENESYS 140 Vis Spectrophotometer with 1 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth5 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 140 Vis Spectrophotometer with 2 year Extended Warranty
Optical Design
Dual beam
Spectral Bandwidth
5 nm
Wavelength Range
325 nm – 1100 nm
Price (USD)
4,580.00
Your Price:
Availability
***
Full specifications
Quantity
TypeGENESYS 140 Vis Spectrophotometer with 2 year Extended Warranty
Wavelength Range325 nm – 1100 nm
DescriptionGENESYS 140 Vis Spectrophotometer with 2 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth5 nm
Unit SizeEach
Catalog Number
Unit Size
Each
Description
GENESYS 140 Vis Spectrophotometer with 3 year Extended Warranty
Optical Design
Dual beam
Spectral Bandwidth
5 nm
Wavelength Range
325 nm – 1100 nm
Price (USD)
4,580.00
Your Price:
Availability
***
Full specifications
Quantity
TypeGENESYS 140 Vis Spectrophotometer with 3 year Extended Warranty
Wavelength Range325 nm – 1100 nm
DescriptionGENESYS 140 Vis Spectrophotometer with 3 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth5 nm
Unit SizeEach
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionOptical DesignSpectral BandwidthWavelength RangePrice (USD)AvailabilityQuantity
840-300000-1YFull specifications
EachGENESYS 150 UV-Vis Spectrophotometer with 1 year Extended Warranty Dual beam2 nm190 nm - 1100 nm
6,450.00
Your Price:
***
TypeGENESYS 150 UV-Vis Spectrophotometer with 1 year Extended Warranty
Wavelength Range190 nm - 1100 nm
DescriptionGENESYS 150 UV-Vis Spectrophotometer with 1 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth2 nm
Unit SizeEach
840-300000Full specifications
EachGENESYS 150 UV-Visible SpectrophotometerDual beam2 nm190 nm – 1100 nm
6,450.00
Your Price:
***
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range190 nm – 1100 nm
Baseline Flatness±0.002A
Compartments
Accommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 150 UV-Visible Spectrophotometer
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0005A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
Lamp Xenon Flash Lamp (>5 years typical, 3 years guaranteed)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display-3A to +5A
Photometric Range–2A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth2 nm
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedSlow, medium and fast - up to 1600 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
840-300300Full specifications
EachGENESYS 150 UV-Vis Spectrophotometer with EU Power CableDual beam2 nm190 nm – 1100 nmRequest A Quote***-
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A±0.004A at 1.0A±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range190 nm – 1100 nm
Baseline Flatness±0.002A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 150 UV-Vis Spectrophotometer with EU Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0005A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–6.0 Hz
KeypadTouchscreen
LampXenon Flash Lamp (>5 years typical, 3 years guaranteed)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display -3A to +5A
Photometric Range–2A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth2 nm
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedSlow, medium and fast - up to 1600 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with 1 year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
840-300400Full specifications
EachGENESYS 150 UV-Vis Spectrophotometer with UK Power CableDual beam2 nm190 nm – 1100 nmRequest A Quote***-
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range190 nm – 1100 nm
Baseline Flatness±0.002A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 150 UV-Vis Spectrophotometer with UK Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0005A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampXenon Flash Lamp (>5 years typical, 3 years guaranteed)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display–3A to +5A
Photometric Range–2A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth2 nm
Stray Light< 1.0%T 198 nm (KCl) , <0.05%T at 220 nm (NaI), <0.03%T at 340 nm (NaNO2)
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
840-308300Full specifications
EachGENESYS 140 Visible Spectrophotometer with EU Power CableDual beam5 nm325 nm – 1100 nmRequest A Quote***-
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-in. color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range325 nm – 1100 nm
Baseline Flatness±0.005A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 140 Visible Spectrophotometer with EU Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5cm
Drift(At 500 nm after 1 hour warmup) <0.0010A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampTungsten-Halogen (>1000 hours)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display–3A to +5A
Photometric Range–3A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth5 nm
Stray Light<0.05%T at 340 nm and 400 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability<±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
840-308000Full specifications
EachGENESYS 140 Visible SpectrophotometerDual beam5 nm325 nm – 1100 nm
4,580.00
Your Price:
***
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-inch color touchscreen, tiltable, high definition, 800 × 1280 pixels
Photometric Accuracy Instrument
±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range325 nm – 1100 nm
Baseline Flatness±0.005A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 140 Visible Spectrophotometer
Dimensions (L x W x H)35.5 x 38.5 x 19.5 cm
Drift(At 500 nm after 1 hour warmup) <0.0010A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampTungsten-Halogen (>1000 hours)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display-3A to +5A
Photometric Range–3A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth5 nm
Stray Light<0.05%T at 340 nm and 400 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability< ±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
 
Unit SizeEach
840-308400Full specifications
EachGENESYS 140 Visible Spectrophotometer with UK Power CableDual beam5 nm325 nm – 1100 nmRequest A Quote***-
Photometric Accuracy Including Standard Tolerance±0.002A at 0.5A
±0.004A at 1.0A
±0.008A at 2.0A
ConnectionsSingle USB-A supports flash memory devices for method and data storage
Duplex USB-A on side supports connection to a Windows™ computer running optional remote control software, keyboard, mouse
Export data to network or PC via USB, Ethernet, or Wi-Fi USB adaptor, available
Print via USB, Ethernet or Wi-Fi USB adaptor available
Detector TypeDual Silicon Photodiodes
Display7-in. color touchscreen, tiltable, high definition, 800 × 1280 pixels
PrinterSnap-on printer available
Wavelength Accuracy±0.5 nm
Wavelength Range325 nm – 1100 nm
Baseline Flatness±0.005A
CompartmentsAccommodates (optional):
8-position cell changer
4-position cell changer (long path cells)
Peltier thermostatted cell holder (20-60 °C)
Sipper accessory
Fiber optic probe coupler
DescriptionGENESYS 140 Visible Spectrophotometer with UK Power Cable
Dimensions (L x W x H)35.5 x 38.5 x 19.5cm
Drift(At 500 nm after 1 hour warmup) <0.0010A/Hr
Electrical RequirementsExternal AC to DC converter. Voltage and Frequency (Hz) selected automatically, 100–240 volts, 50–60 Hz.
KeypadTouchscreen
LampTungsten-Halogen (>1000 hours)
Noise(RMS at 500 nm 60 consecutive measurements)
≤0.00020A at 0A at 260 and 500 nm
≤0.00030A at 1A at 260 and 500 nm
≤0.00040A at 2A at 260 and 500 nm
Optical DesignDual beam
Photometric Display–3A to +5A
Photometric Range–3A to +3.5A
Photometric Repeatability±0.001A at 1A measured at 1.0A at 546 nm
Spectral Bandwidth5 nm
Stray Light<0.05%T at 340 nm and 400 nm
Wavelength Data Interval0.2 nm, 0.5 nm, 1 nm, 2 nm, 5 nm
Wavelength Repeatability<±0.2 nm
Wavelength Scan SpeedAutomatic – up to 1800 nm/min
Weight (Metric)7.5 kg
Warranty2 years standard with one year extension upon registration of the instrument with Thermo Fisher Scientific within 6 months of purchase
Unit SizeEach
840-308000-1YFull specifications
EachGENESYS 140 Vis Spectrophotometer with 1 year Extended Warranty Dual beam5 nm325 nm – 1100 nm
4,580.00
Your Price:
***
TypeGENESYS 140 Vis Spectrophotometer with 1 year Extended Warranty
Wavelength Range325 nm – 1100 nm
DescriptionGENESYS 140 Vis Spectrophotometer with 1 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth5 nm
Unit SizeEach
840-308000-2YFull specifications
EachGENESYS 140 Vis Spectrophotometer with 2 year Extended Warranty Dual beam5 nm325 nm – 1100 nm
4,580.00
Your Price:
***
TypeGENESYS 140 Vis Spectrophotometer with 2 year Extended Warranty
Wavelength Range325 nm – 1100 nm
DescriptionGENESYS 140 Vis Spectrophotometer with 2 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth5 nm
Unit SizeEach
840-308000-3YFull specifications
EachGENESYS 140 Vis Spectrophotometer with 3 year Extended Warranty Dual beam5 nm325 nm – 1100 nm
4,580.00
Your Price:
***
TypeGENESYS 140 Vis Spectrophotometer with 3 year Extended Warranty
Wavelength Range325 nm – 1100 nm
DescriptionGENESYS 140 Vis Spectrophotometer with 3 year Extended Warranty
Optical DesignDual beam
Spectral Bandwidth5 nm
Unit SizeEach
Showing 10 of 12Show more
The GENESYS 150 UV-Vis Spectrophotometer provides automation for high-throughput options and room-light resistance, allowing lid-open operation for improved speed and convenience. A full line of accessories, including automated cell changers and cell holders, sipper, and fiber optic probes are designed to simplify sampling and accommodate high throughput measurements and temperature control. Features data resolution from 0.2nm to 5nm.

The GENESYS 140 Vis Spectrophotometer is a visible spectral region version in the instrumental class. Both instruments feature data resolution from 0.2 nm to 5 nm.
 
Features:
  • 7-inch high-resolution touchscreen user interface
  • Onboard methods, reporting and data export
  • Networking capable (Wi-Fi adaptor available)
  • Room light resistance for "open-lid" operation
  • Sipper accessory
  • Removable, washable sample compartment liner with magnetic placement and hold-down
  • Accommodates cells up to 100 mm pathlength
  • (Optional) 8-position cell changer with switchable turret
  • (Optional) 4-position cell changer (long path cells)
  • (Optional) Peltier-thermostatted single cell holder (20 °C - 60 °C)
 
Designed for:
  • Advanced teaching labs
  • R&D departments
  • Applications requiring high throughput or temperature control

Figures

Frequently asked questions (FAQs)

Citations & References