Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Thermo Scientific™
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Speed up your analysis with high-throughput measurements.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoTypeHolds
840-3034008-position Carousel8 x 10mm Pathlength Cells
840-314700Rect Cell 4-position Carousel4 x 50mm PL Rectangular Cells
840-3036004-position Carousel Base4 x 10mm Pathlength Cells
Número do catálogo 840-303400
Preço (BRL)
-
Type:
8-position Carousel
Holds:
8 x 10mm Pathlength Cells
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.
Especificações
CompatibilityCompatible with 10mm pathlength cells only. Compatible with 4.5mL cell and semi-micro cells with windows ≥ 3mm width (≥ 1000μL). Please note: Not compatible with microcells (less than 1000μL in volume or with GENESYS 30/40/50 spectrophotometers).
For Use With (Equipment)GENESYS 140/150/180, BioMate 160 Spectrophotometers
Holds8 x 10mm Pathlength Cells
Type8-position Carousel
Unit SizeEach