Validation Tools for UV-Visible Spectrophotometers
Actual product may vary
Thermo Scientific™
Validation Tools for UV-Visible Spectrophotometers
Ensure compliance with regulatory guidelines with the Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers providing tools and documentation for system qualification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
840-326000Qualification Filter Kit
840-292900Validator 2 for INSIGHT - US
840-293000Validator 2 for INSIGHT - A4
840-323100GENESYS Validator
840-326100Performance Verification Kit
Catalog number 840-326000
Price (EUR)
-
Type:
Qualification Filter Kit
Get all of the documentation and reference materials needed to facilitate Installation and Operational qualification procedures (IQ/OQ) with Thermo Scientific™ Validation Tools for UV-Visible Spectrophotometers. Certified standards for verifying instrument performance to factory specifications are available separately.
UV-Visible Spectrophotometer Validation Packages provide:
  • Information on the validation process
  • Documentation for manufacturer-verified Installation Qualification (IQ) to ensure proper system performance
  • Information on pharmacopoeia recommended tests for specific instrument performance
  • Installation and instrument certification
Specifications
For Use With (Equipment)Evolution 200 series and Evolution 350 Spectrophotometers
TypeQualification Filter Kit
Unit SizeEach