Evolution™ One Plus USP/EP Standards Pharma Bundle - US - FAQs
View additional product information for Evolution™ One/One Plus UV-Vis Spectrophotometers - FAQs (840-370100, 840-370200, 840-370700, 840-370800, 840-370300, 840-341500, 840-370400, 840-341400, 840-370500, 840-370600)
Error:This product does not have any FAQs available.