MarqMetrix™ Process BallProbe™ Sampling Optic
MarqMetrix™ Process BallProbe™ Sampling Optic
MarqMetrix™ Process BallProbe™ Sampling Optic
Thermo Scientific™

MarqMetrix™ Process BallProbe™ Sampling Optic

The Thermo Scientific™ MarqMetrix™ Process BallProbe™ Sampling Optic is a robust immersion probe for precise, real-time compositional measurements. Its streamlined design and lack of sample preparation requirements allow users to collect reproducible spectra in seconds. Built to withstand tough conditions, it is ideal for various industries where accuracy and reproducibility are crucial. Available in multiple diameters, bundles.
Have Questions?
보기 방식 변경buttonViewtableView
카탈로그 번호프로브 길이포함
840-4008008.5 in. (216 mm)Interchangeable FiberHead, 3/8 in. Process BallProbe
840-40190010 in. (254 mm)1/2 in. Process BallProbe
840-4020008.5 in. (216 mm)3/8 in. Process BallProbe
840-40070010 in. (254 mm)Interchangeable FiberHead, 1/2 in. Process BallProbe
카탈로그 번호 840-400800
제품 가격(KRW)
-
프로브 길이:
8.5 in. (216 mm)
포함:
Interchangeable FiberHead, 3/8 in. Process BallProbe

The Thermo Scientific™ MarqMetrix™ Process BallProbe™ Sampling Optic is an immersion probe for touch measurements in process and laboratory applications. With its streamlined design, users can easily collect reproducible spectra of liquids, solids, slurries, powders, and heterogeneous mixtures in seconds without sample preparation.

Paired with the MarqMetrix All-In-One Process Raman Analyzer, it enables precise, real-time compositional measurements. This corrosion-resistant probe features a gold compression seal and pressure design condition of 6,000 psi, making it suitable for demanding chemical, pressure, and temperature environments. It is ideal for biopharmaceutical, pharmaceutical, food and beverage processing, polymer and plastic manufacturing, and hazardous chemical applications, where measurement accuracy and reproducibility are critical.

The MarqMetrix Process BallProbe Sampling Optic – a robust immersion probe for precise, real-time compositional measurements. Its streamlined design and lack of sample preparation requirements allow users to collect reproducible spectra in seconds. Built to withstand tough conditions, it is ideal for various industries where accuracy and reproducibility are crucial. Available in multiple diameters and bundle sets.

The Thermo Scientific™ MarqMetrix™ Process BallProbe™ Sampling Optic is an immersion probe for touch measurements in process and laboratory applications. With its streamlined design, users can easily collect reproducible spectra of liquids, solids, slurries, powders, and heterogeneous mixtures in seconds without sample preparation.

Paired with the MarqMetrix All-In-One Process Raman Analyzer, it enables precise, real-time compositional measurements. This corrosion-resistant probe features a gold compression seal and pressure design condition of 6,000 psi, making it suitable for demanding chemical, pressure, and temperature environments. It is ideal for biopharmaceutical, pharmaceutical, food and beverage processing, polymer and plastic manufacturing, and hazardous chemical applications, where measurement accuracy and reproducibility are critical.

Features and benefits

  • Immersion probe for touch measurements
  • Measures liquids, solids, slurries, powders, and heterogeneous mixtures
  • Simple design for measurement accuracy and reproducibility
  • Designed to withstand challenging chemical, pressure, and temperature conditions

Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Food and beverage processing
  • Polymer and plastic manufacturing
  • Hazardous chemical applications
  • High pressure and high temperature

Operation conditions

  • Suitable for continuous exposure to dilute and concentrated acids (hot/cold, bases, and most organic solvents, including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane, and acetonitrile
사양
Compatible Wavelengths500 to 1100 nm
설명Interchangeable FiberHead with 3/8 in. Process BallProbe (Bundle set = 3/8in. Process BallProbe + Fiber Head)
용도(장비)MarqMetrix All-In-One Process Raman Analyzer
포함Interchangeable FiberHead, 3/8 in. Process BallProbe
물질Wetted Stainless Steel (C276)
광학 디자인6.00 mm UV-grade sapphire ball
Pressured Design Condition6,000 psi (413 bar)
프로브 길이8.5 in. (216 mm)
Probe Outside Diameter0.375 in. (9.5 mm)
밀봉 재질Gold
온도 범위(영국식 단위)- 20°C to 300°C
디스플레이No Display
제품 유형BallProbe
Unit SizeEach