MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
Thermo Scientific™

MarqMetrix Proximal BallProbe Sampling Optic

The Thermo Scientific™ MarqMetrix™ Proximal BallProbe™ Sampling Optic provides high-signal Raman measurements, offers real-time analysis through barriers, and ideal for a range of applications.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoDescripciónDistancia de trabajo
840-401400Proximal BallProbe de 1/2 in8 mm
840-401500Proximal HV BallProbe100 mm
840-400600Cabezal de fibra intercambiable con Proximal BallProbe8 mm
840-401600Proximal HV BallProbe47 mm
840-400500Cabezal de fibra intercambiable con Proximal HV BallProbe47 mm
840-400400Cabezal de fibra intercambiable con Proximal HV BallProbe100 mm
840-398000Soporte de viales mecanizadoN/D
Número de catálogo 840-401400
Precio (CLP)
-
Descripción:
Proximal BallProbe de 1/2 in
Distancia de trabajo:
8 mm

The Thermo Scientific™ MarqMetrix™ Proximal BallProbe™ Sampling Optic is a high-performance solution for demanding biopharma, pharma, food & beverage, and hazardous chemical applications that require high-signal Raman measurements without physically contacting the sample. Combined with the MarqMetrix All-In-One Process Raman Analyzer, it offers real-time analysis of in-motion samples through physical barriers and air gaps.

The Proximal probe is available in a standard or high-volume (HV) version and bundle sets with a Fiber Head. A vial holder accessory is also available. The HV probe’s long working distance allows it to rapidly analyze in-motion samples on conveyor belts or rotating drums. It is also effective for measuring samples with variable surface relief and uneven packaging. Use it through barriers such as windows, vials, and bags.

Features and benefits

  • Non-contact sampling & analysis

Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Polymer and plastics processing
  • Food and beverage processing
  • Hazardous chemical applications

Operating conditions

  • The MarqMetrix Proximal BallProbe Sampling Optic is intended for use in non-contact sampling but will withstand intermittent immersion in mild chemical solutions
Especificaciones
Compatible Wavelengths500-1100 nm
DescripciónProximal BallProbe de 1/2 in
Para utilizar con (equipo)MarqMetrix All-In-One Process Raman Analyzer
Longitud (imperial)4 in.
Longitud (métrico)100 mm
MaterialOD stainless steel
Diseño ópticoSapphire sampling lens
Probe Outside Diameter.50 in. (12.7 mm)
Material de los cierresHigh-temperature composite adhesive
Intervalo de temperatura (métrico)- 20°C to 100°C
Distancia de trabajo8 mm
PantallaNo Display
Tipo de productoBallProbe
Unit SizeEach