MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
MarqMetrix Proximal BallProbe Sampling Optic
Thermo Scientific™

MarqMetrix Proximal BallProbe Sampling Optic

The Thermo Scientific™ MarqMetrix™ Proximal BallProbe™ Sampling Optic provides high-signal Raman measurements, offers real-time analysis through barriers, and ideal for a range of applications.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoDescriptionWorking Distance
840-401600Proximal HV BallProbe47 mm
840-401400Proximal BallProbe 1/2 in.8 mm
840-401500Proximal HV BallProbe100 mm
840-400600Interchangeable FiberHead w/ Proximal BallProbe8 mm
840-400500Interchangeable FiberHead w/ Proximal HV BallProbe47 mm
840-400400Interchangeable FiberHead w/ Proximal HV BallProbe100 mm
840-398000Machined Vial HolderN/A
Número do catálogo 840-401600
Preço (BRL)
-
Description:
Proximal HV BallProbe
Working Distance:
47 mm

The Thermo Scientific™ MarqMetrix™ Proximal BallProbe™ Sampling Optic is a high-performance solution for demanding biopharma, pharma, food & beverage, and hazardous chemical applications that require high-signal Raman measurements without physically contacting the sample. Combined with the MarqMetrix All-In-One Process Raman Analyzer, it offers real-time analysis of in-motion samples through physical barriers and air gaps.

The Proximal probe is available in a standard or high-volume (HV) version and bundle sets with a Fiber Head. A vial holder accessory is also available. The HV probe’s long working distance allows it to rapidly analyze in-motion samples on conveyor belts or rotating drums. It is also effective for measuring samples with variable surface relief and uneven packaging. Use it through barriers such as windows, vials, and bags.

Features and benefits

  • Non-contact sampling & analysis

Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Polymer and plastics processing
  • Food and beverage processing
  • Hazardous chemical applications

Operating conditions

  • The MarqMetrix Proximal BallProbe Sampling Optic is intended for use in non-contact sampling but will withstand intermittent immersion in mild chemical solutions
Especificações
Compatible Wavelengths500-1100 nm
DescriptionProximal HV BallProbe
For Use With (Equipment)MarqMetrix All-In-One Process Raman Analyzer
Length (English)3 in.
Length (Metric)76.2 mm
MaterialOD stainless steel
Optical Design12.5 mm diameter optical glass
Probe Outside Diameter.56 in. (14.2 mm)
Seal MaterialHigh-temperature composite adhesive
Temperature Range (Metric)- 20°C to 100°C
Working Distance47 mm
DisplayNo Display
Product TypeBallProbe
Unit SizeEach