MarqMetrix Performance BallProbe Sampling Optic
MarqMetrix Performance BallProbe Sampling Optic
MarqMetrix Performance BallProbe Sampling Optic
MarqMetrix Performance BallProbe Sampling Optic
Thermo Scientific™

MarqMetrix Performance BallProbe Sampling Optic

The Thermo Scientific™ MarqMetrix™ Performance BallProbe™ is built for lab and industrial use, offering sensitive TouchRaman Immersion measurements in moderate chemical environments across various sample types.
Have Questions?
Alterar visualizaçãobuttonViewtableView
Número do catálogoProduct TypeOptical Design
840-401800BallProbe3 mm diameter UV-grade sapphire ball
840-401300BallProbe8 mm diameter UV-grade sapphire ball
840-401700BallProbe6 mm diameter UV-grade sapphire ball
840-399700BallProbe with Fiber Head8 mm diameter UV-grade sapphire ball
840-400200BallProbe with Fiber Head6 mm diameter UV-grade sapphire ball
840-400300BallProbe with Fiber Head3.mm diameter UV-grade sapphire ball
Número do catálogo 840-401800
Preço (BRL)
-
Product Type:
BallProbe
Optical Design:
3 mm diameter UV-grade sapphire ball
The Thermo Scientific™MarqMetrix™ Performance BallProbe™ Sampling Optic is designed to withstand moderate chemical environments in laboratory and industrial settings. The standard 0.5 in. diameter MarqMetrix Performance BallProbe Sampling Optic utilizes our largest spherical sapphire ball lens and 316 L stainless steel, enabling highly sensitive TouchRaman Immersion Technology measurements across a wide range of sample types. The MarqMetrix Performance BallProbe Sampling Optic has three standard diameters and chassis length options and is also available in a bundle with an interchangeable fiber head.

Features and benefits

  • Accurate and repeatable measurements
  • Samples liquids, solids, slurries, powders, and heterogeneous mixtures
  • Wide range of chemical compatibility and resistance to high-temperatures
  • Ease of use eliminates user-to-user variation

Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Food and beverage processing
  • Oil and gas processing
  • Polymer and plastic manufacturing
  • Chemical and hazardous applications

Operating conditions

  • Suitable for intermittent exposure to most dilute and concentrated acids (hot and cold), bases, and most organic solvents, including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane, and acetonitrile
  • Avoid exposure to aqua regia, heated formic acid, mixed acid, TFAA
Especificações
Compatible Wavelengths500 to 1100 nm
Description1/8 in. Performance BallProbe (25 mm) Made with high purity UV-grade sapphire ball lens aligned along the C-axis, eliminating response variability due to birefringence. Immersion contact probe for touch measurements in small-volume microwells. It can be used in a moderate chemical laboratory and light-process applications.
For Use With (Equipment)MarqMetrix All-In-One Process Raman Analyzer
MaterialHigh-temperature epoxy (wetted), Stainless Steel (316L)
Optical Design3 mm diameter UV-grade sapphire ball
Pressured Design Condition6000 psi (413 bar)
Probe Length1 in. (25 mm)
Probe Outside Diameter0.0125 in. (3.2 mm)
Temperature Range (Metric)- 20°C to 150°C
DisplayNo Display
Product TypeBallProbe
Unit SizeEach