MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
MarqMetrix FlowCell Sampling Optic
Thermo Scientific™

MarqMetrix FlowCell Sampling Optic

The Thermo Scientific™ MarqMetrix™ FlowCell™ Sampling Optic is for challenging process flow-through applications. This optic ensures robust Raman measurements for your continuous flow process.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionConnections
840-403500Removeable, 4000 psi FlowCell (Gold)Tri-Clamp
840-402700500 psi FlowCell1/8 in. Tube Fitting
840-4028002500 psi FlowCell (Gold)1/8 in. Tube Fitting
840-4030009000 psi FlowCell (Gold)1/4 in. HiP LF4 connections
840-403100Removeable, 4000 psi FlowCell (Gold)1/8 in. Swage
840-403200Removeable, 4000 psi FlowCell (Gold)1/4 Swage
840-403300Removeable, 4000 psi FlowCell (Gold)3/8 Swage
840-403400Removeable, 4000 psi FlowCell (Gold)1/2 Swage
Catalog number 840-403500
Price (CLP)
-
Description:
Removeable, 4000 psi FlowCell (Gold)
Connections:
Tri-Clamp

The Thermo Scientific™ MarqMetrix™ FlowCell™ Sampling Optic is intended for challenging process flow-through applications. The unibody design places the patented BallProbe™ spherical optic directly in the flow path, minimizing dead volume and sample handling requirements while fully utilizing optical efficiencies.

The MarqMetrix FlowCell Sampling Optic is designed for easy integration into continuous flow applications. Plumbing connections are made with standard 1/8 in. tube fitting connectors (e.g., Swagelok, Parker A-lok, Hoke, etc.). Each MarqMetrix FlowCell Sampling Optic is pressure tested so that you can quickly and confidently integrate robust Raman measurements into your continuous flow process.

Features and benefits

  • Ideal for flow-through, continuous flow applications
  • Minimizes dead volume and sample handling requirements
  • Simple design for easy integration
  • Designed to withstand challenging chemical conditions



Applications

  • Biopharmaceutical manufacturing
  • Pharmaceutical manufacturing
  • Food and beverage processing
  • Polymer and plastic manufacturing
  • Hazardous chemical applications



Operating conditions

  • Suitable for continuous exposure to dilute and concentrated acids (hot & cold), bases, and most organic solvents, including ethanol, THF, ethyl acetate, acetone, DCM, toluene, pentane, and acetonitrile
Specifications
Compatible Wavelengths500-1100nm
ConnectionsTri-Clamp
DescriptionRemoveable, 4000 psi FlowCell (Gold)
For Use With (Equipment)MarqMetrix All-In-One Process Raman Analyzer
MaterialWetted = Hastelloy C276
Optical Design6 mm diameter UV-grade sapphire ball BallProbe
Pressured Design Condition
200μL flow path volume 
BallProbe + FlowCell assembly individually pressure tested to 4000 psi (BallProbe pressure design condition is 6000 psi)
Seal MaterialGold
Temperature Range (Metric)- 20°C to 350°C with gold seal
DisplayNo Display
Product TypeBallProbe
Unit SizeEach