Evolution™ Peltier Thermostatting Accessories
Evolution™ Peltier Thermostatting Accessories
Evolution™ Peltier Thermostatting Accessories
Thermo Scientific™
Evolution™ Peltier Thermostatting Accessories
Achieve superior performance in temperature control and sample stirring for many UV-Visible analysis methods.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)TypeIncludes
840-4066006-Cell Rotary PeltierMulti Cell Sample HolderPeltier Cell holder,Water tank
699-131100Evolution spectrophotometersSingle Cell Peltier SystemPeltier Cooling Unit,1-Sample
699-131200Evolution spectrophotometers8-Cell Peltier SystemPeltier Cooling Unit, 8-Sample
840-345000Evolution One/One Plus/ProSingle Cell Peltier SystemPeltier Cell holder,Water tank
840-347500Evolution One/One Plus/ProSingle Cell Peltier SystemKoolance enclosed coolant unit
912A1199Evolution One/One Plus/ProSingle Cell Peltier SystemPeltier Cell holder, Koolance
Catalog number 840-406600
Price (EUR)
-
For Use With (Equipment):
6-Cell Rotary Peltier
Type:
Multi Cell Sample Holder
Includes:
Peltier Cell holder,Water tank
Thermo Scientific™ Peltier Accessories for the Thermo Scientific™ Evolution™ One, Evolution™ One Plus and Evolution™ Pro UV-Vis Spectrophotometers provide thermostatting and ramping with precise temperature control which is important for many UV-Visible analysis methods. Outperforming traditional heater/chiller units, these accessories offer thermostatting and ramping with precise temperature control between 0 °C and 110 °C. Add superior performance with compact integrated design, simple set-up, and fast, precise temperature control.
Precise Thermostatting
Peltier Accessories for the Evolution UV-Visible Spectrophotometers utilize re-circulating water as an effective heat sink to allow the Peltier element to heat or cool the block over a wide temperature range. The single-cell holder can reach as low as 0 °C and as high at 110 °C. Thermo Scientific offers 3 Peltier options to cover many possible applications, including a multi-cell unit. Stirring control for the single cell holders is provided when you place a micro-stir bar inside the cuvette. Unlike thermostatting with a traditional re-circulating water bath where a large volume of liquid must be heated or cooled, heating or cooling the cell holder with the Peltier effect means more rapid and more precise temperature control. Choose between water or chemical coolant to best fit your needs.
Temperature Ramping
Thermo Scientific INSIGHT Pro Software allows you to control and monitor the block temperature of the Peltier unit and allows you to implement programmed temperature ramping with multiple ramp and hold cycles and variable temperature kinetics.  Add a temperature probe hub and probes, and you can monitor the exact temperature in each cell as you perform heating and cooling cycles. The Insight Pro software also includes pre-set methods that can be used to study DNA denaturation and renaturation or other complex temperature programmed experiments.
Interested in instrument and Peltier bundles? Click here for more information.
Specifications
For Use With (Equipment)6-Cell Rotary Peltier
IncludesPeltier Cell holder,Water tank
TypeMulti Cell Sample Holder
Unit SizeEach